Learn More
Biomatik Corporation Human Nadph Oxidase 4 (Nox4) Elisa Kit
Supplier:  Biomatik Corporation EKC34737
Encompass_Preferred
Human Nadph Oxidase 4 (Nox4) Elisa Kit for detecting Nadph Oxidase 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 9.38-600pg/ml. Sensitivity: 2.34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NPH5; NOX4. Target Synonyms: NOX4; RENOX; NADPH oxidase 4; Kidney oxidase-1; KOX-1; Kidney superoxide-producing NADPH oxidase; Renal NAD (PH-oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4160