Learn More
Biomatik Corporation Human Neuritin (Nrn1) Elisa Kit
Supplier:  Biomatik Corporation EKC34774
Encompass_Preferred
Human Neuritin (Nrn1) Elisa Kit for detecting Neuritin 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NPD7; NRN1. Target Synonyms: Neuritin 1; Neuritin; NRN; Nrn1; NRN1_HUMAN; OTTHUMP00000015989; RP3 380B8.2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4165