Learn More
Biomatik Corporation Human Neurturin (Nrtn) Elisa Kit
Supplier:  Biomatik Corporation EKC34813
Encompass_Preferred
Human Neurturin (Nrtn) Elisa Kit for detecting Neurturin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99748; NRTN. Target Synonyms: Neurturin; Neurturin precursor; NRTN; NRTN_HUMAN; NTN; prepro-neurturin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4170