Learn More
Biomatik Corporation Human Nucleolin (Ncl) Elisa Kit
Supplier:  Biomatik Corporation EKC34862
Encompass_Preferred
Human Nucleolin (Ncl) Elisa Kit for detecting Nucleolin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19338; NCL. Target Synonyms: C23; FLJ45706; MS1116; NCL; Nucl; NUCL_HUMAN; Nucleolin; Protein C23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4181