Learn More
Biomatik Corporation Human Nucleoside Diphosphate Kinase A (Nme1) Elisa Kit
Supplier:  Biomatik Corporation EKC34865
Encompass_Preferred
Human Nucleoside Diphosphate Kinase A (Nme1) Elisa Kit for detecting Non-Metastatic Cells 1, Protein (Nm23A) Expressed In in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15531; NME1. Target Synonyms: AWD; AWD; drosophila; homolog of; GAAD; Granzyme A activated DNase; Granzyme A-activated DNase; GZMA activated DNase; Metastasis inhibition factor NM23; NB; NBS; NDK A; NDKA; NDKA_HUMAN; NDP kinase A; NDPK-A; NDPKA; NM23; NM23 long variant; included; nm23-H1; NM23-M1; NM23H1B; included; NME/NM23 nucleoside diphosphate kinase 1; Nme1; NME1-NME2 spliced read-through transcript; included; Non-metastatic cells 1; protein (NM23A) expressed in; Nonmetastatic cells 1; protein expressed in; Nonmetastatic protein 23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4183