Learn More
Biomatik Corporation Human Odontogenic Ameloblast-Associated Protein (Odam) Elisa Kit
Supplier:  Biomatik Corporation EKC34873
Encompass_Preferred
Human Odontogenic Ameloblast-Associated Protein (Odam) Elisa Kit for detecting Odontogenic, Ameloblast Asssociated in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.04ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: A1E959; ODAM. Target Synonyms: ODAM; APIN; Odontogenic ameloblast-associated protein; Apin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4188