Learn More
Biomatik Corporation Human Pappalysin-2(PAPPA2) ELISA kit
Supplier:  Biomatik Corporation EKC34925
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 2 ng/mL-60 ng/mL. Sensitivity: 0.5 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9BXP8. Target Synonyms: RP4-774I24.1, PAPP-A2, PAPPE, PLAC3, placenta-specific 3|pregnancy-associated plasma preproprotein-A2|pregnancy-associated plasma protein-E. Restriction: For Research Use Only.
Catalog No. 50-229-4200