Learn More
Biomatik Corporation Human Pepsinogen Ii Elisa Kit
Supplier:  Biomatik Corporation EKC34942
Encompass_Preferred
Human Pepsinogen Ii Elisa Kit for detecting Pepsinogen Ii in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4202