Learn More
Biomatik Corporation Human phosphoglycerate mutase 2,PGAM2 ELISA Kit
Supplier:  Biomatik Corporation EKC35004
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P15259. Target Synonyms: GSD10, MGC88743, PGAM-M, PGAMM, BPG-dependent PGAM 2|phosphoglycerate mutase 2. Restriction: For Research Use Only.
Catalog No. 50-229-4209