Learn More
Biomatik Corporation Human Plasmin-Antiplasmin Complex, Pap Elisa Kit
Supplier:  Biomatik Corporation EKC35037
Encompass_Preferred
Human Plasmin-Antiplasmin Complex, Pap Elisa Kit for detecting Plasmin-Antiplasmin Complex, Pap in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4216