Learn More
Biomatik Corporation Human Platelet Membrane Glycoprotein Iv, Gp-Iv Elisa Kit
Supplier:  Biomatik Corporation EKC35049
Encompass_Preferred
Human Platelet Membrane Glycoprotein Iv, Gp-Iv Elisa Kit for detecting Cd36 Molecule (Thrombospondin Receptor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.5-160ng/ml. Sensitivity: 0.625ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16671; CD36. Target Synonyms: CD36; GP3B; GP4; Platelet glycoprotein 4; Fatty acid translocase; FAT; Glycoprotein IIIb; GPIIIB; Leukocyte differentiation antigen CD36; PAS IV; PAS-4; Platelet collagen receptor; Platelet glycoprotein IV; GPIV; Thrombospondin receptor; CD antigen CD36. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4220