Learn More
Biomatik Corporation Human Platelet-Derived Growth Factor Cc (Pdgf-Cc) Elisa Kit
Supplier:  Biomatik Corporation EKC35053
Encompass_Preferred
Human Platelet-Derived Growth Factor Cc (Pdgf-Cc) Elisa Kit for detecting Platelet-Derived Growth Factor Cc (Pdgf-Cc) in serum, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4222