Learn More
Biomatik Corporation Human pregnancy associated plasma protein A,PAPP-A ELISA Kit
Supplier:  Biomatik Corporation EKC35080
Encompass_Preferred
Sample Type: serum, plasma, urine. Species Reactivity: Homo sapiens (Human). Detection Range: 1.7 ng/mL-50 ng/mL. Sensitivity: 0.4 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q13219. Target Synonyms: RP11-45A16.1, ASBABP2, DIPLA1, IGFBP-4ase, PAPA, PAPP-A, PAPPA1, aspecific BCL2 ARE-binding protein 2|differentially placenta 1 expressed protein|insulin-like growth factor-dependent IGF binding pro. Restriction: For Research Use Only.
Catalog No. 50-229-4231