Learn More
Biomatik Corporation Human Pro Atrial Natriuretic Peptide, Pro-Anp Elisa Kit
Supplier:  Biomatik Corporation EKC35094
Encompass_Preferred
Human Pro Atrial Natriuretic Peptide, Pro-Anp Elisa Kit for detecting Pro Atrial Natriuretic Peptide, Pro-Anp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4237