Learn More
Biomatik Corporation Human Procollagen Ii N-Terminal Peptide (Piinp) Elisa Kit
Supplier:  Biomatik Corporation EKC35106
Encompass_Preferred
Human Procollagen Ii N-Terminal Peptide (Piinp) Elisa Kit for detecting Procollagen II N-Terminal Peptide(PIInp) in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: procollagen 2 N-terminal peptide (P2NP); procollagen II N-terminal peptide (PIINP). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4239