Learn More
Biomatik Corporation Human Pro-Prostate Specific Antigen (Pro-Psa) Elisa Kit
Supplier:  Biomatik Corporation EKC35148
Encompass_Preferred
Human Pro-Prostate Specific Antigen (Pro-Psa) Elisa Kit for detecting Pro-Prostate Specific Antigen (Pro-Psa) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-4000pg/ml. Sensitivity: 62.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4246