Learn More
Biomatik Corporation Human Protein Dek (Dek) Elisa Kit
Supplier:  Biomatik Corporation EKC35186
Encompass_Preferred
Human Protein Dek (Dek) Elisa Kit for detecting Dek Oncogene in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35659; DEK. Target Synonyms: D6S231E; Dek; DEK gene; DEK oncogene; DEK oncogene DNA binding; DEK_HUMAN; Protein DEK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4252