Learn More
Biomatik Corporation Human Protein Hexim1 (Hexim1) Elisa Kit
Supplier:  Biomatik Corporation EKC35212
Encompass_Preferred
Human Protein Hexim1 (Hexim1) Elisa Kit for detecting Hexamethylene Bis-Acetamide Inducible 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O94992; HEXIM1. Target Synonyms: Cardiac lineage protein 1; CLP 1; CLP1; EDG 1; EDG1; Estrogen down-regulated gene 1 protein; FLJ13562; Hexamethylene bis acetamide inducible 1; Hexamethylene bis acetamide inducible protein; Hexamethylene bis acetamide inducible transcript 1; Hexamethylene bis-acetamide-inducible protein 1; HEXI1_HUMAN; HEXIM 1; Hexim1; HEXIM1 protein; HIS 1; HIS1; HMBA inducible; MAQ 1; MAQ1; Menage a quatre 1; Menage a quatre protein 1; Protein HEXIM1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4263