Learn More
Biomatik Corporation Human Protein Prune Homolog (Prune) Elisa Kit
Supplier:  Biomatik Corporation EKC35228
Encompass_Preferred
Human Protein Prune Homolog (Prune) Elisa Kit for detecting Prune Homolog (Drosophila) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86TP1; PRUNE1. Target Synonyms: DRES-17; DRES17; Drosophila-related expressed sequence 17; hPrune; HTcD37; Protein prune homolog; PRUNE; Prune homolog; PRUNE like protein; PRUNE_HUMAN; TcD37 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4266