Learn More
Biomatik Corporation Human proteinase 3(PR3) ELISA kit
Supplier:  Biomatik Corporation EKC35269
Encompass_Preferred
Sample Type: serum, tissue homogenates, plasma, urine. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78 ng/mL-50 ng/mL. Sensitivity: 0.195 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P24158. Target Synonyms: ACPA, AGP7, C-ANCA, CANCA, MBN, MBT, NP4, P29, PR-3, PR3, Wegener granulomatosis autoantigen|azurophil granule protein 7|leukocyte proteinase 3|myeloblastin|serine proteinase, neutrophil. Restriction: For Research Use Only.
Catalog No. 50-229-4271