Learn More
Biomatik Corporation Human Ras-Related Protein Rab-35 (Rab35) Elisa Kit
Supplier:  Biomatik Corporation EKC35337
Encompass_Preferred
Human Ras-Related Protein Rab-35 (Rab35) Elisa Kit for detecting Rab35, Member Ras Oncogene Family in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15286; RAB35. Target Synonyms: GTP binding protein RAY; GTP-binding protein RAY; H ray; OTTHUMP00000240256; OTTHUMP00000240257; OTTHUMP00000240258; OTTHUMP00000240259; RAB1C; Rab35; RAB35 member RAS oncogene family; RAB35; member RAS oncogene family; RAB35_HUMAN; Ras related protein rab 1c (GTP binding protein ray); Ras related protein Rab 1C; Ras related protein Rab35; Ras-related protein Rab-1C; Ras-related protein Rab-35; RAY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4278