Learn More
Biomatik Corporation Human Regenerating Islet-Derived Protein 3-Gamma (Reg3G) Elisa Kit
Supplier:  Biomatik Corporation EKC35359
Encompass_Preferred
Human Regenerating Islet-Derived Protein 3-Gamma (Reg3G) Elisa Kit for detecting Regenerating Islet-Derived 3 Gamma in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UW15; REG3G. Target Synonyms: LPPM429; Pancreatitis-associated protein 1B; Pancreatitis-associated protein IB; PAP IB; PAP-1B; PAP1B; PAPIB; REG III; Reg III-gamma; REG-3-gamma; REG3G; REG3G_HUMAN; Regenerating islet derived 3 gamma; Regenerating islet-derived protein 3-gamma; Regenerating islet-derived protein III-gamma; UNQ429. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4281