Learn More
Biomatik Corporation Human Secreted Frizzled-Related Protein 3 (Frzb) Elisa Kit
Supplier:  Biomatik Corporation EKC35463
Encompass_Preferred
Human Secreted Frizzled-Related Protein 3 (Frzb) Elisa Kit for detecting Frizzled-Related Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.47-30ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92765; FRZB. Target Synonyms: FRZB; FIZ; FRE; FRP; FRZB1; SFRP3; Secreted frizzled-related protein 3; sFRP-3; Frezzled; Fritz; Frizzled-related protein 1; FrzB-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4291