Learn More
Biomatik Corporation Human Securin (Pttg1) Elisa Kit
Supplier:  Biomatik Corporation EKC35481
Encompass_Preferred
Human Securin (Pttg1) Elisa Kit for detecting Pituitary Tumor-Transforming 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95997; PTTG1. Target Synonyms: AW555095; C87862; Cut2; EAP 1; EAP1; ESP1 associated protein 1; Esp1-associated protein; hPTTG; MGC126883; MGC138276; Pds1; Pituitary tumor transforming 1; Pituitary tumor transforming protein 1; Pituitary tumor-transforming 1; isoform CRA_a; Pituitary tumor-transforming 1; isoform CRA_b; Pituitary tumor-transforming gene 1; Pituitary tumor-transforming gene 1 protein; PTTG 1; PTTG; PTTG1; PTTG1 protein; PTTG1_HUMAN; Pttg3; Securin; Tumor transforming 1; Tumor transforming protein 1; Tumor-transforming protein 1; TUTR 1; TUTR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4294