Learn More
Biomatik Corporation Human Serine/Threonine-Protein Kinase Sgk1 (Sgk1) Elisa Kit
Supplier:  Biomatik Corporation EKC35517
Encompass_Preferred
Human Serine/Threonine-Protein Kinase Sgk1 (Sgk1) Elisa Kit for detecting Serum/Glucocorticoid Regulated Kinase 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O00141; SGK1. Target Synonyms: OTTHUMP00000017247; Serine/threonine protein kinase SGK; Serine/threonine protein kinase Sgk1; Serine/threonine-protein kinase Sgk1; Serum and glucocorticoid regulated kinase; Serum/glucocorticoid regulated kinase 1; Serum/glucocorticoid regulated kinase; Serum/glucocorticoid-regulated kinase 1; SGK 1; SGK; SGK1; Sgk1 variant i3; SGK1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4298