Learn More
Biomatik Corporation Human Serum Amyloid P, Sap Elisa Kit
Supplier:  Biomatik Corporation EKC35542
Encompass_Preferred
Human Serum Amyloid P, Sap Elisa Kit for detecting Amyloid P Component, Serum in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02743; APCS. Target Synonyms: 9.5S alpha 1 glycoprotein; 9.5S alpha-1-glycoprotein; Amyloid P component serum; APCS; MGC88159; Pentaxin related; Pentraxin 2; PTX 2; PTX2; SAMP_HUMAN; SAP; Serum Amyloid P; Serum amyloid P component; Serum amyloid P-component (1-203). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4300