Learn More
Biomatik Corporation Human Stem Cell Factor/Mast Cell Growth Factor, Scf/Mgf Elisa Kit
Supplier:  Biomatik Corporation EKC35667
Encompass_Preferred
Human Stem Cell Factor/Mast Cell Growth Factor, Scf/Mgf Elisa Kit for detecting Kit Ligand in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.45-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21583; KITLG. Target Synonyms: C kit ligand; C-kit ligand; Ckit ligand; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; KIT ligand; Kitl; KITLG; KL 1; KL1; Mast cell growth factor; MGF; MGF stem cell factor; SCF; SCF_HUMAN; SF; SHEP7; sKITLG; Soluble KIT ligand; Steel factor; steel; mouse; homolog of; Stem cell factor; Stem cell factor precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4309