Learn More
Biomatik Corporation Human Sulfhydryl Oxidase 1 (Qsox1) Elisa Kit
Supplier:  Biomatik Corporation EKC35680
Encompass_Preferred
Human Sulfhydryl Oxidase 1 (Qsox1) Elisa Kit for detecting Quiescin Q6 Sulfhydryl Oxidase 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O00391; QSOX1. Target Synonyms: QSOX1; QSCN6; UNQ2520/PRO6013; Sulfhydryl oxidase 1; hQSOX; Quiescin Q6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4311