Learn More
Biomatik Corporation Human Syntaxin-1A(STX1A) ELISA kit
Supplier:  Biomatik Corporation EKC35704
Encompass_Preferred
Sample Type: serum, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q16623. Target Synonyms: HPC-1, STX1, p35-1, syntaxin 1A. Restriction: For Research Use Only.
Catalog No. 50-229-4313