Learn More
Biomatik Corporation Human Thioredoxin-Interacting Protein (Txnip) Elisa Kit
Supplier:  Biomatik Corporation EKC35763
Encompass_Preferred
Human Thioredoxin-Interacting Protein (Txnip) Elisa Kit for detecting Thioredoxin Interacting Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H3M7; TXNIP. Target Synonyms: TXNIP; VDUP1; Thioredoxin-interacting protein; Thioredoxin-binding protein 2; Vitamin D3 up-regulated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4318