Learn More
Biomatik Corporation Human Thyroid-Peroxidase, Tpo Elisa Kit
Supplier:  Biomatik Corporation EKC35788
Encompass_Preferred
Human Thyroid-Peroxidase, Tpo Elisa Kit for detecting Thyroid Peroxidase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07202; TPO. Target Synonyms: MSA; PERT_HUMAN; TDH2A; Thyroid microsomal antigen; Thyroid peroxidase; Thyroperoxidase; TPO; TPX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4321