Learn More
Biomatik Corporation Human Triosephosphate Isomerase (Tpi1) Elisa Kit
Supplier:  Biomatik Corporation EKC35886
Encompass_Preferred
Human Triosephosphate Isomerase (Tpi1) Elisa Kit for detecting Triosephosphate Isomerase 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 28-1800pg/ml. Sensitivity: 7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P60174; TPI1. Target Synonyms: TPI1; TPI; Triosephosphate isomerase; TIM; EC 5.3.1.1; Methylglyoxal synthase; EC 4.2.3.3; Triose-phosphate isomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4327