Learn More
Biomatik Corporation Human Vitamin K-Dependent Protein Z (Proz) Elisa Kit
Supplier:  Biomatik Corporation EKC36037
Encompass_Preferred
Human Vitamin K-Dependent Protein Z (Proz) Elisa Kit for detecting Vitamin K-Dependent Protein Z in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000ng/ml. Sensitivity: 31.25ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22891; PROZ. Target Synonyms: Protein Z; vitamin K dependent plasma glycoprotein; PROZ; PROZ_HUMAN; PZ; Vitamin K-dependent protein Z. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4339