Learn More
Biomatik Corporation Mouse 5-Nucleotidase,5-NT ELISA Kit
Supplier:  Biomatik Corporation EKC36217
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.125 mU/mL-8 mU/mL. Sensitivity: 0.031 mU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q61503. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4358