Learn More
Biomatik Corporation Mouse apolipoprotein H (Apo-H) ELISA Kit
Supplier:  Biomatik Corporation EKC36360
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q01339. Target Synonyms: B2G1, BG, apolipoprotein H. Restriction: For Research Use Only.
Catalog No. 50-229-4367