Learn More
Biomatik Corporation Mouse Carboxyterminal propeptide of type Ⅰ procollagen,PⅠCP ELISA Kit
Supplier:  Biomatik Corporation EKC36470
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Carboxyterminal propeptide of type 1 procollagen,P1CP,_x000D_,Carboxyterminal propeptide of type I procollagen,PICP. Restriction: For Research Use Only.
Catalog No. 50-229-4376