Learn More
Biomatik Corporation Mouse Cd9 Antigen (Cd9) Elisa Kit
Supplier:  Biomatik Corporation EKC36519
Encompass_Preferred
Mouse Cd9 Antigen (Cd9) Elisa Kit for detecting Cd9 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.69-300pg/ml. Sensitivity: 1.17pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40240; Cd9. Target Synonyms: Cd9; CD9 antigen; CD antigen CD9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4381