Learn More
Biomatik Corporation Mouse Coagulation Factor XIII A1 Polypeptide(F13A1)ELISA kit
Supplier:  Biomatik Corporation EKC36579
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8BH61. Target Synonyms: RP11-232H4.1, F13A, FSF, A subunit|TGase|bA525O21.1 (coagulation factor XIII, A1 polypeptide)|coagulation factor XIII A1 subunit|coagulation factor XIII, A polypeptide|factor XIIIa|fibrin stabilizin. Restriction: For Research Use Only.
Catalog No. 50-229-4385