Learn More
Biomatik Corporation Mouse collagen type Ⅱ(Col Ⅱ) antibody ELISA kit
Supplier:  Biomatik Corporation EKC36616
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 U/mL-20 U/mL. Sensitivity: 0.312 U/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: collagen type 2(Col 2) antibody,_x000D_,collagen type II(Col II) antibody. Restriction: For Research Use Only.
Catalog No. 50-229-4387