Learn More
Biomatik Corporation Mouse Cystatin-F(CST7) ELISA kit
Supplier:  Biomatik Corporation EKC36699
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O89098. Target Synonyms: CMAP, cystatin F|cystatin-7|cystatin-F|cystatin-like metastasis-associated protein. Restriction: For Research Use Only.
Catalog No. 50-229-4395