Learn More
Biomatik Corporation Mouse Dickkopf-related protein 3(DKK3) ELISA kit
Supplier:  Biomatik Corporation EKC36737
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 9.4 pg/mL-600 pg/mL. Sensitivity: 2.35 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9QUN9. Target Synonyms: REIC, RIG, RIG-like 5-6|RIG-like 7-1|dickkopf 3|dickkopf homolog 3|regulated in glioma. Restriction: For Research Use Only.
Catalog No. 50-229-4398