Learn More
Biomatik Corporation Mouse Factor-related Apoptosis ligand,FASL ELISA Kit
Supplier:  Biomatik Corporation EKC36839
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P41047. Target Synonyms: APT1LG1, CD178, CD95L, FASL, TNFSF6, CD95 ligand|apoptosis (APO-1) antigen ligand 1|fas ligand|tumor necrosis factor (ligand) superfamily, member 6. Restriction: For Research Use Only.
Catalog No. 50-229-4405