Learn More
Biomatik Corporation Mouse Fatty acid synthase(FASN) ELISA kit
Catalog No. 502294407
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P19096. Target Synonyms: FAS, MGC14367, MGC15706, OA-519, SDR27X1, short chain dehydrogenase/reductase family 27X, member 1. Restriction: For Research Use Only.
This item is not returnable. View return policy
Catalog No. 50-229-4407 Supplier Biomatik Corporation Supplier No. EKC36841
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P19096. Target Synonyms: FAS, MGC14367, MGC15706, OA-519, SDR27X1, short chain dehydrogenase/reductase family 27X, member 1. Restriction: For Research Use Only.