Learn More
Biomatik Corporation Mouse Gelsolin Elisa Kit
Supplier:  Biomatik Corporation EKC36937
Encompass_Preferred
Mouse Gelsolin Elisa Kit for detecting Gelsolin (Amyloidosis, Finnish Type) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13020; Gsn. Target Synonyms: Gsn; GsbGelsolin; Actin-depolymerizing factor; ADF; Brevin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4412