Learn More
Biomatik Corporation Mouse Glutaredoxin-Related Protein 5, Mitochondrial (Glrx5) Elisa Kit
Supplier:  Biomatik Corporation EKC36962
Encompass_Preferred
Mouse Glutaredoxin-Related Protein 5, Mitochondrial (Glrx5) Elisa Kit for detecting Glutaredoxin 5 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q80Y14; Glrx5. Target Synonyms: Glrx5Glutaredoxin-related protein 5; mitochondrial; Monothiol glutaredoxin-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4415