Learn More
Biomatik Corporation Mouse guanylate cyclase activator 2B (uroguanylin) (GUCA2B) ELISA kit
Catalog No. 502294418
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 ng/ml-4000ng/ml. Sensitivity: 15.6 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O09051. Target Synonyms: Not Available. Restriction: For Research Use Only.
This item is not returnable. View return policy
Catalog No. 50-229-4418 Supplier Biomatik Corporation Supplier No. EKC37014
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 ng/ml-4000ng/ml. Sensitivity: 15.6 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O09051. Target Synonyms: Not Available. Restriction: For Research Use Only.