Learn More
Biomatik Corporation Mouse Membrane primary amine oxidase(AOC3) ELISA kit
Supplier:  Biomatik Corporation EKC37380
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.25 ng/mL-80 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O70423. Target Synonyms: HPAO, SSAO, VAP-1, VAP1, amine oxidase, copper containing 3|copper amine oxidase|semicarbazide-sensitive amine oxidase|vascular adhesion protein 1. Restriction: For Research Use Only.
Catalog No. 50-229-4440