Learn More
Biomatik Corporation Mouse nuclear factor-κB p65,NF-κB p65 ELISA Kit
Supplier:  Biomatik Corporation EKC37511
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q04207. Target Synonyms: MGC131774, NFKB3, p65, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3|v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor of kappa light polypeptide gene. Restriction: For Research Use Only.
Catalog No. 50-229-4446