Learn More
Biomatik Corporation Mouse Oxyntomodulin Elisa Kit
Supplier:  Biomatik Corporation EKC37535
Encompass_Preferred
Mouse Oxyntomodulin Elisa Kit for detecting Oxyntomodulin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20pg/ml. Sensitivity: 0.078pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4449