Learn More
Biomatik Corporation Mouse Platelet-Derived Growth Factor Cc (Pdgf-Cc) Elisa Kit
Supplier:  Biomatik Corporation EKC37597
Encompass_Preferred
Mouse Platelet-Derived Growth Factor Cc (Pdgf-Cc) Elisa Kit for detecting Platelet Derived Growth Factor C in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8CI19; Pdgfc. Target Synonyms: Pdgfc; Scdgf; Platelet-derived growth factor C; PDGF-C; Fallotein; Spinal cord-derived growth factor; SCDGF; VEGF-E) [Cleaved into: Platelet-derived growth factor C; latent form; PDGFC latent form); Platelet-derived growth factor C; receptor-binding form; PDGFC receptor-binding form)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4455